Supelco® Organic Reference Standard, BTEX Solution
Fournisseur: Merck
About this item
- GHS signal: GHS08
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501
2 Options Available Below
Product Details & Documents
- GHS signal: GHS08
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 43728-1ML
- 43728-10X1ML
- M-Clarity quality level:
- MQ100
- MQ100
- Density:
- 1,099 g/ml at 20 °C
- 1,099 g/ml at 20 °C
- Hazard class description:
- H340 - H350
- H340 - H350
- Flash point:
- Not applicable
- Not applicable
- Shipping conditions:
- STD
- STD
- Storage conditions:
- 2...8 °C
- 2...8 °C
Specifications
Product Family Options
Product Information
- StandardsPkDisponibilitéPrix
- 43728-1MLBTEX SolutionSpecifications:
Plus
Specifications
Cat. No.43728-1MLM-Clarity quality levelMQ100Density1,099 g/ml at 20 °CHazard class descriptionH340 - H350Flash pointNot applicableShipping conditionsSTDStorage conditions2...8 °C - 43728-10X1MLBTEX SolutionSpecifications:
Plus
Specifications
Cat. No.43728-10X1MLM-Clarity quality levelMQ100Density1,099 g/ml at 20 °CHazard class descriptionH340 - H350Flash pointNot applicableShipping conditionsSTDStorage conditions2...8 °C



