Order Entry
Belgium
ContactUsLinkComponent
Mono-(2-Ethyl-5-hydroxyhexyl) phthalate, mixture of diastereomers, analytical standard, Supelco®
Mono-(2-Ethyl-5-hydroxyhexyl) phthalate, mixture of diastereomers, analytical standard, Supelco®
Catalog # 09171-50MG
Supplier:  Merck
CAS Number:  
undefined
Mono-(2-Ethyl-5-hydroxyhexyl) phthalate, mixture of diastereomers, analytical standard, Supelco®
Catalog # 09171-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • CAS No.:
    40321-99-1
  • Formula:
    C₁₆H₂₂O₅
  • Storage temperature:
    2…8 °C
  • Description:
    mono-(2-Ethyl-5-hydroxyhexyl) phthalate, ≥97,0% (HPLC)

Specifications

About this item