Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Aroclor spike mix, analytical standard, Supelco®
Aroclor spike mix, analytical standard, Supelco®
Aroclor spike mix, analytical standard, Supelco®
Catalog # 861274.
Supplier:
Merck
CAS Number:
Expand image
Aroclor spike mix, analytical standard, Supelco®
Catalog # 861274.
Supplier:
Merck
CAS Number:
Specifications
Concentration:
100 mug/mL each component in acetone
Pk:
1
Description:
Aroclor Spike Mix
Type:
Analytical standard
Specifications
About this item
Product Details
Product Details