Order Entry
Belgium
ContactUsLinkComponent
EPA TCL Pesticides mix, certified reference material, Supelco®
EPA TCL Pesticides mix, certified reference material, Supelco®
Catalog # 48913.
Supplier:  Merck
CAS Number:  
EPA TCL Pesticides mix, certified reference material, Supelco®
Catalog # 48913.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in hexane: toluene (1:1)
  • Pk:
    1
  • Description:
    EPA TCL Pesticides Mix
  • Type:
    Certified reference material

Specifications

About this item