Order Entry
Belgium
ContactUsLinkComponent
 
Semivolatile Base/Neutrals Surrogate Spike Mix, certified reference material, Supelco®
Catalog # 47262.
Supplier:  Merck
CAS Number:  
Semivolatile Base/Neutrals Surrogate Spike Mix, certified reference material, Supelco®
Catalog # 47262.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5000 mug/mL each component in methylene chloride
  • Pk:
    1
  • Description:
    Semivolatile Base/Neutrals Surrogate Spike Mix
  • Type:
    Certified reference material

Specifications

About this item