Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Indoor Air Standard
Supelco® Organic Reference Standard, Indoor Air Standard
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Indoor Air Standard
Supplier:  Merck

Specifications

  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Description:
    4M9148-U
  • Flash point:
    9,7 °C - Closed cup

Specifications

Frequently Bought Together