Supelco® Organic Reference Standard, EPA 8260 Internal Standards Mix HC
Catalog # 47082-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:10000 μg/ml in methanol
- Pk:1
- Standards:Certified reference material
- Description:EPA 8260 Internal standards mix HC
- Storage conditions:2…8 °C
- Hazard class description:H226 - H301 + H311 + H331 - H351 - H370 - H411
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:50 °C
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311