ContactUsLinkComponent
EPA 8260 volatiles calibration mix, certified reference material, Supelco®
  500607.
 :  Merck
 :  
EPA 8260 volatiles calibration mix, certified reference material, Supelco®
  500607.
 :  Merck
 :  
  • Concentration:
    2000 mug/mL each component in methanol
  • Pk:
    1
  • Description:
    EPA 8260 Volatiles Calibration Mix
  • Type:
    Certified reference material