Supelco® PESTANAL® Organic Reference Standard, Thiacloprid-(Thiazolidin Ring-D₄)
Catalog # 30673-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- Description:Thiacloprid-(thiazolidin ring-d₄)
- Storage conditions:2...30 °C
- Hazard class description:H301 - H332 - H336 - H351 - H360FD - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Thiacloprid-d₄; [3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidin-d₄-ylidene]cyanamide
- Formula:C₁₀D₄H₅ClN₄S
- Molecular weight:256,75
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P202 - P261 - P264 - P273 - P301 + P310 - P304 + P340 + P312