Order Entry
Belgium
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Terbuthylazine-(ethyl-d₅)
  91799-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Terbuthylazine-(ethyl-d₅)
  91799-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    222986-60-9
  • Description:
    Terbuthylazine-(ethyl-d₅)
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01632311
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N²-tert-Butyl-6-chloro-N⁴-ethyl-d₅-1,3,5-triazine-2,4-diamine; Terbuthylazine-d₅
  • Molecular weight:
    234,74

 

 

  • GHS signal: GHS07, GHS09
 : P273 - P301 + P312 + P330