Supelco® PESTANAL® Organic Reference Standard, TDCPP
32951-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:13674-87-8
- Description:TDCPP
- Storage conditions:2...30 °C
- Hazard class description:H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD00083121
- M-Clarity quality level:MQ100
- Synonym(s):Tris(1,3-dichloro-2-propyl) phosphate; 1,3-Dichloro-2-propanol phosphate; Phosphoric acid tris(1,3-dichloro-2-propyl ester)
- Formula:C₉H₁₅Cl₆O₄P
- Molecular weight:430,90
- Density:1,514 g/ml at 20 °C (lit.)
- Flash point:250 - 252 °C - open cup
- GHS signal: GHS08, GHS09
: P202 - P273 - P280 - P308 + P313 - P391 - P405