Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Spirodiclofen
Supelco® PESTANAL® Organic Reference Standard, Spirodiclofen
Catalog # 33654-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Spirodiclofen
Catalog # 33654-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    148477-71-8
  • Description:
    Spirodiclofen
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H317 - H350 - H361f - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04972571
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl) 2,2-dimethylbutyrate
  • Formula:
    C₂₁H₂₄Cl₂O₄
  • Molecular weight:
    411,32
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P202 - P260 - P273 - P280 - P302 + P352 - P308 + P313