Pesticide reference standard, Simazine hydroxy, analytical standard, Supelco®
Catalog # MET58B
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:2599-11-3
- Description:Simazine hydroxy
- Formula:C₇H₁₃N₅O
- Description:Simazine hydroxy