Supelco® PESTANAL® Organic Reference Standard, Phenthoate
31611-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2597-03-7
- Description:Phenthoate
- Storage conditions:2...8 °C
- Hazard class description:H301 + H311 - H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00145099
- M-Clarity quality level:MQ100
- Formula:C₁₂H₁₇O₄PS₂
- Molecular weight:320,36
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310