Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Oleamide
Supelco® Organic Reference Standard, Oleamide
Catalog # 08393-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Oleamide
Catalog # 08393-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    301-02-0
  • Description:
    Oleamide
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H413
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00053638
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    cis-9,10-Octadecenoamide; cis-9-Octadecenamide; Oleic acid amide
  • Formula:
    C₁₈H₃₅NO
  • Molecular weight:
    281,48
  • Storage temperature:
    –20 °C
  • Flash point:
    207 °C

Specifications

About this item

  • GHS signal: GHS07
Caution: P273 - P501