Supelco® Organic Reference Standard, Monononadecanoin
Catalog # 02193-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:112340-30-4
- Description:Monononadecanoin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Glyceryl monononadecanoate
- Formula:C₂₂H₄₄O₄
- Molecular weight:372,58
- Flash point:Not applicable