Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Methapyrilene HCl, analytical standard, Supelco®
Organic reference standard, Methapyrilene HCl, analytical standard, Supelco®
Catalog # 442641.
Supplier:  Merck
CAS Number:  
Organic reference standard, Methapyrilene HCl, analytical standard, Supelco®
Catalog # 442641.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • CAS No.:
    135-23-9
  • Description:
    Methapyrilene HCl
  • Formula:
    C₁₄H₂₀ClN₃S
  • Description:
    Methapyrilene HCl

Specifications

About this item