Supelco® PESTANAL® Organic Reference Standard, Lactofen
Catalog # 32972-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:77501-63-4
- Description:Lactofen
- Storage conditions:2...30 °C
- Hazard class description:H312 - H411
- Shipping conditions:STD
- MDL No.:MFCD01632761
- M-Clarity quality level:MQ100
- Synonym(s):2-Ethoxy-1-methyl-2-oxoethyl 5-(2-chloro-4-trifluoromethylphenoxy)-2-nitrobenzoate; Ethyl O-[5-(2-chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoyl]-ᴅʟ-lactate
- Formula:C₁₉H₁₅ClF₃NO₇
- Molecular weight:461,77
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P280 - P302 + P352 + P312