Supelco® PESTANAL® Organic Reference Standard, Isoproturon-D₆
34017-10MG-R
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:217487-17-7
- Description:Isoproturon-d₆
- Storage conditions:2...30 °C
- Hazard class description:H351 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD01632296
- M-Clarity quality level:MQ100
- Synonym(s):3-(4-Isopropylphenyl)-1,1-dimethylurea-d₆
- Formula:C₁₂H₁₂D₆N₂O
- Molecular weight:212,32
- Flash point:Not applicable
- GHS signal: GHS08, GHS07, GHS09
: P202 - P260 - P273 - P280 - P308 + P313 - P391