Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Imidacloprid-Olefin
Supelco® PESTANAL® Organic Reference Standard, Imidacloprid-Olefin
Catalog # 34534-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Imidacloprid-Olefin
Catalog # 34534-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    115086-54-9
  • Description:
    Imidacloprid-olefin
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD02660798
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-1H-imidazol-2-amine
  • Formula:
    C₉H₈ClN₅O₂
  • Molecular weight:
    253,65
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P280 - P301 + P312 + P330 - P304 + P340 + P312 - P305 + P351 + P338 - P337 + P313