Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Hexachlorobenzene-¹³C₆
Supelco® PESTANAL® Organic Reference Standard, Hexachlorobenzene-¹³C₆
Catalog # 68696-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Hexachlorobenzene-¹³C₆
Catalog # 68696-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    93952-14-8
  • Description:
    Hexachlorobenzene-¹³C₆
  • Storage conditions:
    RT
  • Hazard class description:
    H350 - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144438
  • M-Clarity quality level:
    MQ200
  • Formula:
    ¹³C₆Cl₆
  • Molecular weight:
    290,74

Specifications

About this item

  • GHS signal: GHS08, GHS09
Caution: P201 - P202 - P260 - P264 - P273 - P308 + P313