Supelco® PESTANAL® Organic Reference Standard, Forchlorfenuron
Catalog # 32974-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:68157-60-8
- Description:Forchlorfenuron
- Storage conditions:2...30 °C
- Hazard class description:H351 - H411
- Shipping conditions:STD
- MDL No.:MFCD00059898
- M-Clarity quality level:MQ100
- Synonym(s):N-(2-Chloro-4-pyridyl)-N′-phenylurea; 4-CPPU; CPPU
- Formula:C₁₂H₁₀ClN₃O
- Molecular weight:247,68
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405