Pesticide reference standard, Fluchloralin, PESTANAL®, analytical standard, Supelco®
Catalog # 45500-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:33245-39-5
- Description:Fluchloralin
- M-Clarity quality level:MQ100
- Formula:C₁₂H₁₃ClF₃N₃O₄
- Description:Fluchloralin