Supelco® PESTANAL® Organic Reference Standard, Flubenzimin
Catalog # 45499-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:37893-02-0
- Description:Flubenzimin
- Storage conditions:2...30 °C
- Hazard class description:H319 - H331 - H410
- Shipping conditions:STD
- MDL No.:MFCD00078680
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₀F₆N₄S
- Molecular weight:416,34
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P264 - P271 - P273 - P304 + P340 + P311 - P305 + P351 + P338