Supelco® PESTANAL® Organic Reference Standard, Fluazifop-P
Catalog # 35372-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:83066-88-0
- Description:Fluazifop-P
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD20036281
- M-Clarity quality level:MQ100
- Synonym(s):(R)-2-[4-(5-Trifluoromethyl-2-pyridyloxy)phenoxy]propionic acid
- Formula:C₁₅H₁₂F₃NO₄
- Molecular weight:327,26
- Flash point:Not applicable