Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fentrazamide
Supelco® PESTANAL® Organic Reference Standard, Fentrazamide
Catalog # 37903-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fentrazamide
Catalog # 37903-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    158237-07-1
  • Description:
    Fentrazamide
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04972563
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-(2-Chlorophenyl)-N-cyclohexyl-N-ethyl-4,5-dihydro-5-oxo-1H-tetrazole-1-carboxamide
  • Formula:
    C₁₆H₂₀ClN₅O₂
  • Molecular weight:
    349,82
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273