Supelco® PESTANAL® Organic Reference Standard, Fensulfothion
45307-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:115-90-2
- Description:Fensulfothion
- Storage conditions:2...8 °C
- Hazard class description:H300 + H310 + H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055487
- M-Clarity quality level:MQ100
- Synonym(s):Diethyl 4-(methylthio)phenyl phosphate
- Formula:C₁₁H₁₇O₄PS₂
- Molecular weight:308,35
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P262 - P264 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310