Supelco® PESTANAL® Organic Reference Standard, Fenpiclonil
36532-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:74738-17-3
- Description:Fenpiclonil
- Storage conditions:2...30 °C
- Hazard class description:H332
- Shipping conditions:STD
- MDL No.:MFCD00144304
- M-Clarity quality level:MQ100
- Formula:C₁₁H₆Cl₂N₂
- Molecular weight:237,08
- Flash point:Not applicable
- GHS signal: GHS07
: P261 - P271 - P304 + P340 + P312