Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fenamidone
Supelco® PESTANAL® Organic Reference Standard, Fenamidone
Catalog # 33965-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fenamidone
Catalog # 33965-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    161326-34-7
  • Description:
    Fenamidone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H312 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03792823
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (S)-1-Anilino-4-methyl-2-methylthio-4-phenyl-2-imidazolin-5-one
  • Formula:
    C₁₇H₁₇N₃OS
  • Molecular weight:
    311,40
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P280 - P302 + P352 + P312 - P362 + P364 - P391 - P501