Supelco® PESTANAL® Organic Reference Standard, Desnitro-Imidacloprid Hydrochloride
Catalog # 37052-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:127202-53-3
- Description:Desnitro-imidacloprid hydrochloride
- Storage conditions:2...30 °C
- Hazard class description:H302 - H373
- Shipping conditions:STD
- MDL No.:MFCD22373625
- M-Clarity quality level:MQ100
- Synonym(s):1-(2-Chloro-pyridin-5-yl-methyl)-2-imino-imidazolidine hydrochloride
- Formula:C₂₄H₃₂O₆
- Molecular weight:247,12
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS07
Caution: P260 - P264 - P270 - P301 + P312 - P314 - P501