Supelco® PESTANAL® Organic Reference Standard, Clofentezine
Catalog # 36763-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:74115-24-5
- Description:Clofentezine
- Storage conditions:2...30 °C
- Hazard class description:H312 - H410
- Shipping conditions:STD
- MDL No.:MFCD00143998
- M-Clarity quality level:MQ100
- Synonym(s):3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine; Clofentezine
- Melting point:182...185 °C (lit.)
- Formula:C₁₄H₈Cl₂N₄
- Molecular weight:303,15
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P280 - P302 + P352 + P312 - P362 + P364 - P391 - P501