Supelco® PESTANAL® Organic Reference Standard, Chlorbensid
Catalog # 36123-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:103-17-3
- Description:Chlorbensid
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00055267
- M-Clarity quality level:MQ100
- Synonym(s):4-Chlorobenzyl 4-chlorophenyl sulfide
- Formula:C₁₃H₁₀Cl₂S
- Molecular weight:269,19
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07