Order Entry
Belgium
ContactUsLinkComponent
 
Pesticide reference standard, Bitertanol, PESTANAL®, analytical standard, Supelco®
Catalog # 45349-250MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Bitertanol, PESTANAL®, analytical standard, Supelco®
Catalog # 45349-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2114693
  • Description:
    Bitertanol
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₀H₂₃N₃O₂
  • Description:
    Bitertanol

Specifications

About this item