Order Entry
Belgium
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Bistrifluron
Catalog # 65563-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Bistrifluron
Catalog # 65563-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    201593-84-2
  • Description:
    Bistrifluron
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD22581210
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-[2-Chloro-3,5-bis(trifluoromethyl)phenyl]-3-(2,6-difluorobenzoyl)urea
  • Molecular weight:
    446,68

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P501