Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, BDE No 1
Supelco® Organic Reference Standard, BDE No 1
Catalog # 43691-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, BDE No 1
Catalog # 43691-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    7025-06-1
  • Description:
    BDE No 1
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00626462
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Bromo-2-phenoxybenzene; 2-Bromodiphenyl ether; PBDE-1
  • Formula:
    C₁₂H₉BrO
  • Molecular weight:
    249,10
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P301 + P312 + P330