Supelco® PESTANAL® Organic Reference Standard, Azoxystrobin
31697-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:131860-33-8
- Description:Azoxystrobin
- Storage conditions:2...30 °C
- Hazard class description:H331 - H410
- Shipping conditions:STD
- MDL No.:MFCD08277047
- M-Clarity quality level:MQ100
- Formula:C₂₂H₁₇N₃O₅
- Molecular weight:403,39
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P261 - P271 - P273 - P304 + P340 + P311 - P391 - P403 + P233