Supelco® PESTANAL® Organic Reference Standard, Acetamiprid-N-Desmethyl
Catalog # 32979-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:190604-92-3
- Description:Acetamiprid-N-desmethyl
- Storage conditions:2...30 °C
- Hazard class description:H301 - H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD08690484
- M-Clarity quality level:MQ100
- Synonym(s):N-(6-Chloro-3-pyridylmethyl)-N′-cyano-acetamidine
- Formula:C₉H₉ClN₄
- Molecular weight:208,65
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Caution: P301 + P310 + P330 - P302 + P352 - P305 + P351 + P338