Supelco® PESTANAL® Organic Reference Standard, 5-Hydroxythiabendazole
Catalog # 33818-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:948-71-0
- Description:5-Hydroxythiabendazole
- Storage conditions:2...30 °C
- Hazard class description:H302 - H400
- Shipping conditions:STD
- MDL No.:MFCD00152219
- M-Clarity quality level:MQ100
- Synonym(s):2-(4-Thiazolyl)-5-benzimidazolol; 5-Hydroxy-2-(4-thiazolyl)benzimidazole
- Formula:C₁₀H₇N₃OS
- Molecular weight:217,25
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P301 + P312 + P330