Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Palmitoylglycerol
Supelco® Organic Reference Standard, 2-Palmitoylglycerol
Catalog # 75614-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 2-Palmitoylglycerol
Catalog # 75614-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    23470-00-0
  • Description:
    2-Palmitoylglycerol
  • Storage conditions:
    –70 °C
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00058513
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    β-Monopalmitin; 2-Hexadecanoylglycerol; 2-Monohexadecanoylglycerol; 2-Monopalmitin; 2-Monopalmitoylglycerol; Hexadecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester
  • Molecular weight:
    330,50

Specifications

About this item