Orders LinkContactUsLinkComponent
Poly(α-methylstyrene) analytical standards, for GPC
Poly(α-methylstyrene) analytical standards, for GPC
Supplier:   Merck
Poly(α-methylstyrene) analytical standards, for GPC
Supplier:  Merck
Certificates
Poly(α-methylstyrene) analytical standards, for GPC
Poly(α-methylstyrene) analytical standards, for GPC
3 Options Available Below
Product Details & Documents
Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.
Certifications: Exact values can be taken from the accompanying certificate of analysis.
Caution: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 81516-250MG
  • 81520-250MG
  • 81523-250MG
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 81516-250MG
    ~7800 Mn, Mw/Mn ~1,07
    ~7800 Mn, Mw/Mn ~1,07
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg
    Specifications:
    More
    Specifications
    Cat. No.
    81516-250MG
    M-Clarity quality level
    MQ100
  • 81520-250MG
    ~106000 Mn, Mw/Mn ~1,02
    ~106000 Mn, Mw/Mn ~1,02
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 161,00 €
    161,00 €
    1 * 250 mg
       161,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    81520-250MG
    M-Clarity quality level
    MQ100
  • 81523-250MG
    ~427000 Mn, Mw/Mn ~1,02
    ~427000 Mn, Mw/Mn ~1,02
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg
    Specifications:
    More
    Specifications
    Cat. No.
    81523-250MG
    M-Clarity quality level
    MQ100