ContactUsLinkComponent
Microscope clamp, gooseneck, Dino-Lite RK-02
Microscope clamp, gooseneck, Dino-Lite RK-02
Catalog # IDCPRK-02
undefined
Microscope clamp, gooseneck, Dino-Lite RK-02
Catalog # IDCPRK-02
Specifications
  • Spanning width:
    70 mm
  • Description:
    Gooseneck clamp for Dino-Lite microscopes, RK-02, with flexible arm (60 cm), can be clamped to surfaces up to 7 cm thick
  • Seq:
    916
  • Pk:
    1
Specifications
About this item
The RK-02 is a durable gooseneck clamp designed for flexible and versatile usage with any Dino-Lite digital microscope.
  • C-clamp fits to virtually any surface up to 7 cm (~3 inch) thick
  • Can be mounted horizontally or vertically
With this sturdy stand, users can easily position the Dino-Lite by bending the 60 cm long arm and adjusting the rotating head. The base of the RK-02 can also be rotated providing even more freedom and it can be fixed with the quick release fastener for use on uneven surfaces.
Compatible with all Dino-Lite models, except the high magnification models.