ContactUsLinkComponent
Three-fold multi-tap bank, Discofix® C
Three-fold multi-tap bank, Discofix® C
Catalog # BRAU16615C
Supplier:  B. Braun
undefined
Three-fold multi-tap bank, Discofix® C
Catalog # BRAU16615C
Supplier:  B. Braun
Specifications
  • Colour:
    Blue
  • Description:
    Three fold tap banks, drug-resistant
  • Length:
    150 cm
  • Pk:
    100
Specifications
About this item
Drug-Resistant Multi-Tap Systems are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
  • Leaks caused by stress cracks, contamination and air embolism are prevented
  • Easy access through the rotary adapter for injections and aspirations
  • Control of the 45° position by 'clicking'