Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite solution 13% (w/v), Technical Grade
Corrosive
Environment
Sodium hypochlorite solution 13% (w/v), Technical Grade
Supplier:  PanReac AppliChem
Certificates
Corrosive
Environment
Sodium hypochlorite solution 13% (w/v), Technical Grade
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (Active Chlorine, Iodometry):
    10.0 - 14.0%
  • NaCl:
    Max. 11.5%

Specification Test Results

Frequently Bought Together