Order Entry
Belgium
ContactUsLinkComponent
Hemacolor® Rapid staining of blood smear solution I fixative reagent, Sigma-Aldrich®
Flammable
Toxic
Health
Hemacolor® Rapid staining of blood smear solution I fixative reagent, Sigma-Aldrich®
Catalog # 1.11955.2500
Supplier:  Merck
CAS Number:  
undefined
Flammable
Toxic
Health
Hemacolor® Rapid staining of blood smear solution I fixative reagent, Sigma-Aldrich®
Catalog # 1.11955.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Pk Info:
    Preparation Capacity 1250 - 5000 slides

Specifications

About this item

 :
UN:1230
ADR:3 (6.1),
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
 :1