Order Entry
Belgium
ContactUsLinkComponent
Tuerk solution stain for hematology, Sigma-Aldrich®
Tuerk solution stain for hematology, Sigma-Aldrich®
Catalog # 1.09277.0100
Supplier:  Merck
CAS Number:  
Tuerk solution stain for hematology, Sigma-Aldrich®
Catalog # 1.09277.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

About this item

 :
Density:1.00 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
 :1
MDL Number:MFCD00148719