Order Entry
Belgium
ContactUsLinkComponent
Contamination control floor mats, Protectamat
Contamination control floor mats, Protectamat
Catalog # DYCECC02/06/1BLAU
Supplier:  DYCEM
Contamination control floor mats, Protectamat
Catalog # DYCECC02/06/1BLAU
Supplier:  DYCEM

Specifications

  • Type:
    With red safety leaders
  • Colour:
    Blue
  • L×W:
    2000×4000 mm
  • Thickness:
    7 mm
  • Pk:
    1

Specifications

About this item

This ProtectaMat is a high performance contamination control zone which effectively attracts, collects and retains foot and wheel borne contamination, preventing particles and microbes from getting into and out of critical areas.

  • Quick and easy to maintain
  • Ready for immediate use
  • Complete with red safety leaders to minimise safety hazards
  • Non toxic with neutral ESD features

This mat prevents over 99% of viable and non-viable contamination from entering the critical area.

Heat resistance working range: 0 to 50 °C

Load resistance: 40 kg/cm (570 lbs./in²)