Order Entry
Belgium
ContactUsLinkComponent
 
Flammable
Irritant
Health
Carbon disulphide ≥99.5% (by GC) for synthesis
Catalog # APPC161244.1611
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Flammable
Irritant
Health
Carbon disulphide ≥99.5% (by GC) for synthesis
Catalog # APPC161244.1611
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

Specification Test Results

  • Minimum Assay (GC):
    ≤99.5%
  • Identity (IR):
    Passes Test
  • Water (H₂O):
    ≤0.05%

Specification Test Results

About this item

 :
 :76.14 g/mol
Boiling Pt:46 °C
Melting Pt:–111 °C
Density:1.264 g/cm³
EINECS:200-843-6
UN:1131
ADR:3 (6.1),I
 :GHS02
 :GHS07
 :GHS08
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321