Orders LinkContactUsLinkComponent
Precision™ General purpose water baths
Precision™ General purpose water baths
Supplier:   THERMO ELECTRON LED
Precision™ General purpose water baths
Certificates
Precision™ General purpose water baths
Precision™ General purpose water baths
6 Options Available Below
Product Details & Documents
Precision™ general purpose water baths are designed to maintain water temperature from ambient to 100 °C.
  • Smaller footprint, compared to previous models, frees up valuable benchtop space
  • Advanced microprocessor controller is designed for extended functionality
  • Protects work with audible alarms
  • Conveniently save commonly used settings with four temperature presets
These rugged, high-performance baths range from 2 to 28 litres, including shallow models. Over-temperature safety circuitry is designed to prevent thermal runaway, while new auto-on and auto-off timers allow you to optimize operation schedules. Benefit from outstanding chemical and corrosion resistance with epoxy powder-coated exterior, easily clean the chamber with its seamless stainless steel interior.
The temperature uniformity lies at a range of ±0,2 °C.
Electrical information: 100 to 115 V/200 to 230 V, 50/60 Hz
Certifications: cULus Listed, CE compliant, US FDA Class I Medical Device.
Delivery information: Baths come with clear polycarbonate gable cover, diffuser tray, drain hose and rubber duck. Includes a setting for thermal beads. Comes with N5-15 Plug.
Documents
Product Family Options
Product Information
  • Temperature Range
    Temp. stability
    Capacity
    Bath W×D×H
    W×D×H
    Availability
    Price
  • HAKATSGP02
    General purpose water bath, GP 02, 200 W
    General purpose water bath, GP 02, 200 W
    Ambient...90 °C
    ±0,1 °C
    2 L
    138×155×150 mm
    230×199×233 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 975,00 €
    975,00 €
    1 * 1 ST
       975,00 €
  • HAKATSGP05
    General purpose water bath, GP 05, 300 W
    General purpose water bath, GP 05, 300 W
    Ambient...100 °C
    ±0,1 °C
    5 L
    154×300×150 mm
    246×355×232 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 080,00 €
    1 080,00 €
    1 * 1 ST
       1 080,00 €
  • HAKATSGP10
    General purpose water bath, GP 10, 800 W
    General purpose water bath, GP 10, 800 W
    Ambient...100 °C
    ±0,1 °C
    10 L
    300×153×65 mm
    355×246×232 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €
  • HAKATSGP15D
    General purpose water bath, GP15D, 300 W and 800 W
    General purpose water bath, GP15D, 300 W and 800 W
    Ambient...100 °C
    ±0,1 °C
    5 L and 10 L (Dual)
    154×300×150 mm and 301×330×150 mm
    392×587×233 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 220,00 €
    2 220,00 €
    1 * 1 ST
       2 220,00 €
  • HAKATSGP20
    General purpose water bath, GP 20, 1200 W
    General purpose water bath, GP 20, 1200 W
    Ambient...100 °C
    ±0,1 °C
    20 L
    297×500×150 mm
    392×555×233 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €
  • HAKATSGP28
    General purpose water bath, GP 28, 1200 W
    General purpose water bath, GP 28, 1200 W
    Ambient...100 °C
    ±0,1 °C
    28 L
    297×500×200 mm
    392×555×282 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 660,00 €
    1 660,00 €
    1 * 1 ST
       1 660,00 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Concentric ring cover, 4 holes
    Concentric ring cover, 4 holes
    HAKA1546230Q
    GP10
    Log in for availability
    1 * 1 ST - 824,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Concentric ring cover, 6 holes
    Concentric ring cover, 6 holes
    HAKA1546231Q
    GP20, GP28
    Log in for availability
    1 * 1 ST - 1 060,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Polycarbonate gable cover
    Polycarbonate gable cover
    HAKATSGPACL05
    GP05, GP2S
    Log in for availability
    1 * 1 ST - 253,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Polycarbonate gable cover
    Polycarbonate gable cover
    HAKATSGPACL10
    GP10
    Log in for availability
    1 * 1 ST - 291,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Polycarbonate gable cover
    Polycarbonate gable cover
    HAKATSGPACL20
    GP20, GP28
    Log in for availability
    1 * 1 ST - 320,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack
    Stainless steel test tube rack
    HAKA3161601
    GP20, GP28
    Log in for availability
    1 * 1 ST - 548,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel gable cover
    Stainless steel gable cover
    HAKAGPSSL20
    GP20, GP28
    Log in for availability
    1 * 1 ST - 511,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Polycarbonate gable cover
    Polycarbonate gable cover
    HAKATSGPACL02
    GP02
    Log in for availability
    1 * 1 ST - 222,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel Petri dish rack
    Stainless steel Petri dish rack
    HAKA3166183
    GP20, GP28
    Log in for availability
    1 * 1 ST - 431,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel gable cover
    Stainless steel gable cover
    HAKAGPSSL02
    GP02
    Log in for availability
    1 * 1 ST - 421,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel gable cover
    Stainless steel gable cover
    HAKAGPSSL05
    GP05, GP2S
    Log in for availability
    1 * 1 ST - 462,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel gable cover
    Stainless steel gable cover
    HAKAGPSSL10
    GP10
    Log in for availability
    1 * 1 ST - 474,00 €