Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dropping pipettes

Dropping pipettes

Micropipettes, 100 µl, 83 mm
Catalog # DELT201110
Supplier:   DELTALAB
Dropping pipettes
Catalog # DELT201110
Supplier:  DELTALAB
Dropping pipettes

Micropipettes, 100 µl, 83 mm

Price
100,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Made of transparent, inert LDPE.

  • Can be frozen in liquid nitrogen
  • Volumetric tolerance: ±10%

Ideal for use as volume measurers, to incorporate in kits of reagents, pregnancy tests.

Press the upper bulb before use, filling all the stem, and the rest remains in the inferior bulb.

Documents

Specifications
  • Capacity:
    100 µl
  • Description:
    Micropipettes
  • Length:
    83 mm