Orders LinkContactUsLinkComponent
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
Supplier:  LIOFILCHEM
Certificates
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
15 Options Available Below
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Product Family Options
Product Information
  • Availability
    Price
  • 85615.
    ESC swab Buffered Peptone Water + Neutralising, 100 tubes×4 ml
    ESC swab Buffered Peptone Water + Neutralising, 100 tubes×4 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 111,00 €
    111,00 €
    1 * 100 ST
       111,00 €
  • 85603.
    ESC swab Buffered Peptone Water, 100 tubes×10 ml
    ESC swab Buffered Peptone Water, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85604.
    ESC swab Buffered Peptone Water, 100 tubes×5 ml
    ESC swab Buffered Peptone Water, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85605.
    ESC swab D/E Neutralising Broth, 100 tubes×10 ml
    ESC swab D/E Neutralising Broth, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85606.
    ESC swab D/E Neutralising Broth, 100 tubes×5 ml
    ESC swab D/E Neutralising Broth, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85614.
    ESC swab Demi Fraser Broth, 100 tubes×10 ml
    ESC swab Demi Fraser Broth, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85607.
    ESC swab Letheen Broth, 100 tubes×10 ml
    ESC swab Letheen Broth, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85608.
    ESC swab Letheen Broth, 100 tubes×5 ml
    ESC swab Letheen Broth, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85621.
    ESC swab Letheen Broth, 100 tubes×1 ml
    ESC swab Letheen Broth, 100 tubes×1 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 109,00 €
    109,00 €
    1 * 100 ST
       109,00 €
  • 85609.
    ESC swab Maximum Recovery Diluent, 100 tubes×10 ml
    ESC swab Maximum Recovery Diluent, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85610.
    ESC swab Maximum Recovery Diluent, 100 tubes×5 ml
    ESC swab Maximum Recovery Diluent, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85612.
    ESC swab Mycoplasma Transport Broth, 100 tubes×5 ml
    ESC swab Mycoplasma Transport Broth, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85601.
    ESC swab Neutralising Rinse Solution, 100 tubes×10 ml
    ESC swab Neutralising Rinse Solution, 100 tubes×10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 139,00 €
    139,00 €
    1 * 100 ST
       139,00 €
  • 85602.
    ESC swab Neutralising Rinse Solution, 100 tubes×5 ml
    ESC swab Neutralising Rinse Solution, 100 tubes×5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
  • 85623.
    ESC swab Buffered Peptone Water + Neutralising, 100 tubes×1 ml
    ESC swab Buffered Peptone Water + Neutralising, 100 tubes×1 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 109,00 €
    109,00 €
    1 * 100 ST
       109,00 €